1,3-Di-tert-butylbenzol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

272051-1G
SIAL272051-1GEA 114 EUR
SIAL272051-1G
1,3-Di-tert-butylbenzol, Sigma-Aldrich®
1,3-Di-tert-butylbenzol
Formel: C₁₄H₂₂
Molecular Weight: 190,33 g/mol
MDL: MFCD00008830
CAS-Nummer: 1014-60-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR