(4-Brombutyl)triphenylphosphoniumbromid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

272132-25G
SIAL272132-25GEA 94.3 EUR
SIAL272132-25G
(4-Brombutyl)triphenylphosphoniumbromid, Sigma-Aldrich®
(4-Bromobutyl)triphenylphosphonium bromide
Formel: C₂₂H₂₃Br₂P
Molecular Weight: 478,21 g/mol
Schmelzpunkt: 215 °C
MDL: MFCD00031636
CAS-Nummer: 7333-63-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR