5-Methylchinoxalin, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ100
272310-1G
SIAL272310-1GEA
61.8
EUR
SIAL272310-1G
5-Methylchinoxalin, Sigma-Aldrich®
5-Methylquinoxaline
Formel:
C₉H₈N₂ Molecular Weight: 144,18 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00012335 CAS-Nummer: 13708-12-8 EINECS: 237-246-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...