2,4,6-Triphenylpyriliumtetrafluoroborat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,4,6-Triphenylpyrylium tetrafluoroborate

M-Clarity™ Qualitätsstufe = MQ100

272345-5G
SIAL272345-5GEA 81.4 EUR
SIAL272345-5G
2,4,6-Triphenylpyriliumtetrafluoroborat, Sigma-Aldrich®
2,4,6-Triphenylpyriliumtetrafluoroborat
Formel: C₂₃H₁₇BF₄O
Molecular Weight: 396,19 g/mol
MDL: MFCD00012001
CAS-Nummer: 448-61-3
EINECS: 207-186-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR