Hafnium, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

272558-1EA
SIAL272558-1EAEA 433 EUR
SIAL272558-1EA
Hafnium, Sigma-Aldrich®
Hafnium
Formel: Hf
MDL: MFCD00011032
CAS-Nummer: 7440-58-6
EINECS: 231-166-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR