Benzoylacetonitril, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-Phenyl-3-ketopropionsäurenitril, 3-Phenyl-3-oxopropionsäurenitril
M-Clarity™ Qualitätsstufe = MQ100
272728-5G
SIAL272728-5GEA
42.8
EUR
SIAL272728-5G
Benzoylacetonitril, Sigma-Aldrich®
Benzoylacetonitril
Formel:
(C₆H₅)C(O)CH₂CN Molecular Weight: 145,16 g/mol Siedepunkt: 301 °C (1013 hPa) Schmelzpunkt: 83,5 °C |
MDL:
MFCD00001942 CAS-Nummer: 614-16-4 EINECS: 210-369-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...