Arsen(III)ethanolat, Sigma-Aldrich®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

272744-5G
SIAL272744-5GEA 419 EUR
SIAL272744-5G
Arsen(III)ethanolat, Sigma-Aldrich®
Arsen(III)ethanolat
Formel: C₆H₁₅AsO₃
MDL: MFCD00015120
CAS-Nummer: 3141-12-6
EINECS: 221-543-1
UN: 2929
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR