1,3-Di-tert-butyltoluol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,3-Di-tert-butyl-5-methylbenzene

M-Clarity™ Qualitätsstufe = MQ100

273015-25G
SIAL273015-25GEA 83 EUR
SIAL273015-25G
1,3-Di-tert-butyltoluol, Sigma-Aldrich®
1,3-Di-tert-butyltoluol
Formel: C₁₅H₂₄
Molecular Weight: 204,36 g/mol
MDL: MFCD00026300
CAS-Nummer: 15181-11-0
EINECS: 239-230-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR