3,5-Di-tert-butylbenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

273023-5G
SIAL273023-5GEA 136 EUR
SIAL273023-5G
3,5-Di-tert-butylbenzoesäure, Sigma-Aldrich®
3,5-Di-tert-butylbenzoesäure
Formel: C₁₅H₂₂O₂
Molecular Weight: 234,34 g/mol
MDL: MFCD00082727
CAS-Nummer: 16225-26-6
EINECS: 240-350-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR