3-Chloroperoxybenzoic acid ≤77% (durch titrimetrische Analyse)

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ200

273031-100G 273031-25G 273031-500G 273031-1KG
SIAL273031-100GEA 100 EUR
SIAL273031-100G SIAL273031-25G SIAL273031-500G SIAL273031-1KG
3-Chloroperoxybenzoic acid ≤77% (durch titrimetrische Analyse)
3-Chloroperoxybenzoic acid

3-Chloroperbenzoic acid has been used in the oxidation of N-p-tolylsulfinyl-(E)-1-phenylethylideneimine to N-p-tolylsulfonyl (E)-1-phenylethylideneimine using dichloromethane as a solvent.


Used in the preparation of mono- and di-epoxy-functionalised poly(3-hydroxybutyrate)-based reactive polymers. MCPBA is also an effective oxidizing agent for conversion of α-amide substituted polyesters to their corresponding polysulfones with tunable thermal properties.


3-Chloroperbenzoic acid is a strong oxidising agent mainly used for the epoxidation of alkenes and also in the Baeyer-Villiger oxidation of ketones to esters.

Formel: C₇H₅ClO₃
Lagertemperatur: Kühlschrank
MDL: MFCD00002127
CAS-Nummer: 937-14-4
UN: 3106
ADR: 5.2,

Bestellen

Spezifikation Testergebnisse

Proton NMR Spectrum Conforms to Structure
Assay ≤77%
Impurity, 3-Chlorobenzoic Acid ≥6.0%
Titration by Na₂S₂O₃ ≤77.0%
Water (by Karl Fischer) ≥17.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR