3,5-Di-tert-butylanilin, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

273104-1G
SIAL273104-1GEA 70 EUR
SIAL273104-1G
3,5-Di-tert-butylanilin, Sigma-Aldrich®
3,5-Di-tert-butylanilin
Formel: C₁₄H₂₃N
Molecular Weight: 205,34 g/mol
MDL: MFCD00134096
CAS-Nummer: 2380-36-1
EINECS: 219-173-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR