Methyl-4-indolcarboxylat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

273880-1G
SIAL273880-1GEA 27.1 EUR
SIAL273880-1G
Methyl-4-indolcarboxylat, Sigma-Aldrich®
Methyl-4-indolcarboxylat
Formel: C₁₀H₉NO₂
Molecular Weight: 175,19 g/mol
MDL: MFCD00191222
CAS-Nummer: 39830-66-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR