6,7-Dimethoxy-1-tetralon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6,7-Dimethoxy-3,4-dihydronaphthalen-1(2H)-one

M-Clarity™ Qualitätsstufe = MQ100

273937-5G
SIAL273937-5GEA 246 EUR
SIAL273937-5G
6,7-Dimethoxy-1-tetralon, Sigma-Aldrich®
6,7-Dimethoxy-1-tetralon
Formel: C₁₂H₁₄O₃
Molecular Weight: 206,24 g/mol
MDL: MFCD00134100
CAS-Nummer: 13575-75-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR