ω-Brom-2-nitroacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Brommethyl-o-nitrophenylketon, 2-Brom-2'-nitroacetophenon, α-Brom-2-nitroacetophenon, 2-Nitrophenacylbromid

274542-5G
SIAL274542-5GEA 265 EUR
SIAL274542-5G
ω-Brom-2-nitroacetophenon, Sigma-Aldrich®
ω-Brom-2-nitroacetophenon
Formel: C₈H₆BrNO₃
Molecular Weight: 244,04 g/mol
MDL: MFCD00010294
CAS-Nummer: 6851-99-6
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR