(±)-Citronellal
Lieferant: Merck
Synonyme:
(±)-3,7-Dimethyl-6-octenal
27470-100ML-F
SIAL27470-100ML-FEA
66.8
EUR
SIAL27470-100ML-F
(±)-Citronellal
(±)-Citronellal
Formel:
C₁₀H₁₈O Molecular Weight: 154,25 g/mol Siedepunkt: 207 °C (1013 hPa) Schmelzpunkt: –15 °C Dichte: 0,8548 g/cm³ (20 °C) |
MDL:
MFCD00038090 CAS-Nummer: 106-23-0 EINECS: 203-376-6 Merck Index: 13,02353 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...