4-Nitrophthalsäure, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
4-Nitro-1,2-benzoldicarbonsäure
274755-25G
SIAL274755-25GEA
73.1
EUR
SIAL274755-25G
4-Nitrophthalsäure, Sigma-Aldrich®
4-Nitrophthalsäure
Formel:
C₈H₅NO₆ Schmelzpunkt: 162…165 °C |
MDL:
MFCD00007252 CAS-Nummer: 610-27-5 EINECS: 210-215-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...