D(-)-Citrullin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: H-D-Cit-OH

27505-500MG-F
SIAL27505-500MG-FEA 196 EUR
SIAL27505-500MG-F
D(-)-Citrullin, Sigma-Aldrich®
D(-)-Citrullin
Formel: C₆H₁₃N₃O₃
Molecular Weight: 175,19 g/mol
MDL: MFCD00216469
CAS-Nummer: 13594-51-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR