cis-Bis(benzonitril)dichlorplatin(II), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

275816-500MG
SIAL275816-500MGEA 139 EUR
SIAL275816-500MG
cis-Bis(benzonitril)dichlorplatin(II), Sigma-Aldrich®
cis-Bis(benzonitril)dichlorplatin(II)
Formel: C₁₄H₁₀Cl₂N₂Pt
MDL: MFCD00013125
CAS-Nummer: 15617-19-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR