Methyl-3-aminopyrazin-2-carboxylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 2-aminopyrazine-3-carboxylate

M-Clarity™ Qualitätsstufe = MQ100

276154-10G
SIAL276154-10GEA 48.8 EUR
SIAL276154-10G
Methyl-3-aminopyrazin-2-carboxylat, Sigma-Aldrich®
Methyl-3-aminopyrazin-2-carboxylat
Formel: C₆H₇N₃O₂
MDL: MFCD00010097
CAS-Nummer: 16298-03-6
EINECS: 240-387-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR