Acetonitril-[2-13C] (99% ¹³C ), Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
277223-250MG
SIAL277223-250MGEA
443
EUR
SIAL277223-250MG
Acetonitril-[2-13C] (99% ¹³C ), Sigma-Aldrich®
Acetonitril-[2-13C]
Formel:
C₂H₃N |
MDL:
MFCD00001879 CAS-Nummer: 1722-09-4 UN: 1648 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...