Ölsäure Ph. Eur., Sigma-Aldrich®
Lieferant: Merck
Synonyme:
cis-9-Octadecensäure
M-Clarity™ Qualitätsstufe = MQ200
Oleic acid is a fatty acid. It activates protein kinase C in hepatocytes and also inhibits 2,4-dinitrophenol-stimulated ATPase.
- Assay (GC): 65,0 - 88,0%
- Form: liquid (clear)
- Density: 0,89 g/ml at 25 °C (lit.)
Oleic acid is used as a surfactant for the stabilization of magnetic nanoparticles synthesized by the traditional coprecipitation method and in the preparation of 10-hydroxyoctadecanoic acid and methyl ester of 10-hydroxyoctadecanoic acid.
Standards: Meets analytical specification of Ph. Eur.
Formel:
C₁₈H₃₄O₂ Molecular Weight: 282.47 g/mol Siedepunkt: 286 Schmelzpunkt: 16,7 Flammpunkt: 180 |
MDL:
MFCD00064242 CAS-Nummer: 112-80-1 Merck Index: 13,06898 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...