Ölsäure Ph. Eur., Sigma-Aldrich®

Lieferant: Merck

Synonyme: cis-9-Octadecensäure

M-Clarity™ Qualitätsstufe = MQ200

27728-1L-R 27728-2.5L-R 27728-25KG-R
SIAL27728-1L-REA 112 EUR
SIAL27728-1L-R SIAL27728-2.5L-R SIAL27728-25KG-R
Ölsäure Ph. Eur., Sigma-Aldrich®
Ölsäure

Oleic acid is a fatty acid. It activates protein kinase C in hepatocytes and also inhibits 2,4-dinitrophenol-stimulated ATPase.


  • Assay (GC): 65,0 - 88,0%
  • Form: liquid (clear)
  • Density: 0,89 g/ml at 25 °C (lit.)


Oleic acid is used as a surfactant for the stabilization of magnetic nanoparticles synthesized by the traditional coprecipitation method and in the preparation of 10-hydroxyoctadecanoic acid and methyl ester of 10-hydroxyoctadecanoic acid.


Standards: Meets analytical specification of Ph. Eur.

Formel: C₁₈H₃₄O₂
Molecular Weight: 282.47 g/mol
Siedepunkt: 286
Schmelzpunkt: 16,7
Flammpunkt: 180
MDL: MFCD00064242
CAS-Nummer: 112-80-1
Merck Index: 13,06898
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5aecc5dce4d0318386a45d7adb7c5822631010a&vid=892a2df4ecab3679b06983b84cd27ad334ef961f&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR