4,5-Dichlorimidazol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4,5-Dichlor-1H-imidazol
M-Clarity™ Qualitätsstufe = MQ100
277363-5G
SIAL277363-5GEA
62.4
EUR
SIAL277363-5G
4,5-Dichlorimidazol, Sigma-Aldrich®
4,5-Dichloroimidazole
Formel:
C₃H₂Cl₂N₂ Molecular Weight: 136,97 g/mol |
MDL:
MFCD00005195 CAS-Nummer: 15965-30-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...