(±)-Methyl-2-methylbutyrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl2-methylbutyrate

M-Clarity™ Qualitätsstufe = MQ100

277452-50ML
SIAL277452-50MLEA 77.9 EUR
SIAL277452-50ML
(±)-Methyl-2-methylbutyrat, Sigma-Aldrich®
(±)-Methyl-2-methylbutyrat
Formel: C₆H₁₂O₂
Molecular Weight: 116,16 g/mol
MDL: MFCD00009335
CAS-Nummer: 868-57-5
EINECS: 212-778-0
UN: 3272
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR