(±)-2-Methyltetrahydrofuran-3-on, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Methyltetrahydrofuran-3-one

M-Clarity™ Qualitätsstufe = MQ100

277487-5G
SIAL277487-5GEA 22.6 EUR
SIAL277487-5G
(±)-2-Methyltetrahydrofuran-3-on, Sigma-Aldrich®
(±)-2-Methyltetrahydrofuran-3-on
Formel: C₅H₈O₂
Molecular Weight: 100,12 g/mol
MDL: MFCD00010423
CAS-Nummer: 3188-00-9
EINECS: 221-685-4
UN: 1224
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR