2-Octinsäuremethylester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl-2-octinoat

M-Clarity™ Qualitätsstufe = MQ100

277509-25ML
SIAL277509-25MLEA 135 EUR
SIAL277509-25ML
2-Octinsäuremethylester, Sigma-Aldrich®
2-Octynoic acid methyl ester
Formel: C₉H₁₄O₂
Molecular Weight: 154,21 g/mol
MDL: MFCD00009530
CAS-Nummer: 111-12-6
EINECS: 203-836-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR