(2E,6E)-3,7,11-Trimethyl-2,6,10-dodecatrien-1-ol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2E,6E)-3,7,11-Trimethyldodeca-2,6,10-trien-1-ol

M-Clarity™ Qualitätsstufe = MQ200

277541-10G 277541-1G
SIAL277541-10GEA 204 EUR
SIAL277541-10G SIAL277541-1G
(2E,6E)-3,7,11-Trimethyl-2,6,10-dodecatrien-1-ol, Sigma-Aldrich®
trans,trans-Farnesol
Formel: C₁₅H₂₆O
Molecular Weight: 222.37 g/mol
MDL: MFCD00002918
CAS-Nummer: 106-28-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR