3-Methyl-3-oxetanmethanol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

277681-10G 277681-50G
SIAL277681-10GEA 49.2 EUR
SIAL277681-10G SIAL277681-50G
3-Methyl-3-oxetanmethanol, Sigma-Aldrich®
3-Methyl-3-oxetanmethanol
Formel: C₅H₁₀O₂
MDL: MFCD00010273
CAS-Nummer: 3143-02-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR