Diethyl-tert-butylmalonat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

278041-5G
SIAL278041-5GEA 137 EUR
SIAL278041-5G
Diethyl-tert-butylmalonat, Sigma-Aldrich®
Diethyl-tert-butylmalonat
Formel: C₁₁H₂₀O₄
Molecular Weight: 216,28 g/mol
MDL: MFCD00009152
CAS-Nummer: 759-24-0
EINECS: 212-067-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR