(R)-(+)-Menthyltoluol-4-sulfinat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (1S,2R,5S)-(+)-Menthyl (R)-p-Toluenesulphinate

M-Clarity™ Qualitätsstufe = MQ200

278742-10G
SIAL278742-10GEA 301 EUR
SIAL278742-10G
(R)-(+)-Menthyltoluol-4-sulfinat, Sigma-Aldrich®
(R)-(+)-Menthyltoluol-4-sulfinat
Formel: C₁₇H₂₆O₂S
MDL: MFCD00064472
CAS-Nummer: 91796-57-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR