(S)-(-)-Menthyltoluol-4-sulfinat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (1R,2S,5R)-(-)-Menthyl (S)-p-toluenesulphinate, (S)-((1R,2S,5R)-2-isopropyl-5-methylcyclohexyl) 4-methylbenzenesulphinate

M-Clarity™ Qualitätsstufe = MQ200

278750-10G
SIAL278750-10GEA 156 EUR
SIAL278750-10G
(S)-(-)-Menthyltoluol-4-sulfinat, Sigma-Aldrich®
(S)-(-)-Menthyltoluol-4-sulfinat
Formel: C₁₇H₂₆O₂S
Molecular Weight: 294,4521 g/mol
MDL: MFCD00010192
CAS-Nummer: 1517-82-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR