Iod(V)oxid, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ100
278890-100G
278890-25G
SIAL278890-100GEA
283
EUR
SIAL278890-100G
SIAL278890-25G
Iod(V)oxid, Sigma-Aldrich®
Iod(V)oxid
Formel:
I₂O₅ Dichte: 4,79 g/cm³ (20 °C) |
MDL:
MFCD00011356 CAS-Nummer: 12029-98-0 EINECS: 234-740-2 UN: 1479 ADR: 5.1,III Merck Index: 12,05040 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...