1-(4-Nitrophenyl)piperazin, Sigma-Aldrich®

Lieferant: Merck

279773-5G
SIAL279773-5GEA 52.8 EUR
SIAL279773-5G
1-(4-Nitrophenyl)piperazin, Sigma-Aldrich®
1-(4-Nitrophenyl)piperazin
Formel: C₁₀H₁₃N₃O₂
Schmelzpunkt: 136,85 °C
MDL: MFCD00005961
CAS-Nummer: 6269-89-2
EINECS: 228-443-7
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR