5-Methyl-1,3-cyclohexandion, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 5-Methylcyclohexane-1,3-dione

M-Clarity™ Qualitätsstufe = MQ100

279951-5G
SIAL279951-5GEA 178 EUR
SIAL279951-5G
5-Methyl-1,3-cyclohexandion, Sigma-Aldrich®
5-Methyl-1,3-cyclohexandion
Formel: C₇H₁₀O₂
Molecular Weight: 126,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00010379
CAS-Nummer: 4341-24-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR