1-Butylpyrrolidin, Sigma-Aldrich®

Lieferant: Merck

280372-100ML
SIAL280372-100MLEA 347 EUR
SIAL280372-100ML
1-Butylpyrrolidin, Sigma-Aldrich®
1-Butylpyrrolidin
Formel: C₈H₁₇N
Molecular Weight: 127,23 g/mol
CAS-Nummer: 767-10-2
EINECS: 212-179-4
UN: 2929
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR