Tantalcarbid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tantalum(IV) carbide

280801-10G
SIAL280801-10GEA 41.8 EUR
SIAL280801-10G
Tantalcarbid, Sigma-Aldrich®
Tantalum carbide
Formel: CTa
Molecular Weight: 192.96 g/mol
Siedepunkt: 5500
Schmelzpunkt: 3880
MDL: MFCD00011255
CAS-Nummer: 12070-06-3
EINECS: 235-118-3
UN: 3178
ADR: 4.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR