Tetrabutylammoniumtetraphenylborat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tetrabutylammonium Tetraphenylborate

M-Clarity™ Qualitätsstufe = MQ100

281034-5G
SIAL281034-5GEA 160 EUR
SIAL281034-5G
Tetrabutylammoniumtetraphenylborat, Sigma-Aldrich®
Tetrabutylammonium tetraphenylborate
Formel: C₄₀H₅₆BN
Molecular Weight: 561,6904 g/mol
MDL: MFCD00011749
CAS-Nummer: 15522-59-5
EINECS: 239-562-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR