Kaliumtetracyanoplatinat(II), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

281557-1G
SIAL281557-1GEA 193 EUR
SIAL281557-1G
Kaliumtetracyanoplatinat(II), Sigma-Aldrich®
Potassium tetracyanoplatinate(II)
Formel: C₄K₂N₄Pt
Molecular Weight: 377,34 g/mol
MDL: MFCD00011374
CAS-Nummer: 562-76-5
EINECS: 209-236-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR