Potassium tetracyanoplatinate(II) hydrate ≥98%, kristallin, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: Potassium platinum(II) cyanide hydrate

M-Clarity™ Qualitätsstufe = MQ100

281565-1G
SIAL281565-1GEA 207 EUR
SIAL281565-1G
Potassium tetracyanoplatinate(II) hydrate ≥98%, kristallin, Sigma-Aldrich®
Potassium tetracyanoplatinate(II) hydrate
Formel: K₂Pt(CN)₄·xH₂O
Molecular Weight: 377,34 g/mol (anhy)
MDL: MFCD00149920
CAS-Nummer: 38245-27-1
EINECS: 209-236-0
UN: 2811
ADR: 6.1,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR