trans-4-Nitrozimtaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: trans-3-(4-Nitrophenyl)-2-propenal, (E)-3-(4-Nitrophenyl)acrylaldehyde, (E)-4-Nitrocinnamaldehyde, (E)-3-(4-Nitrophenyl)-2-propenal

M-Clarity™ Qualitätsstufe = MQ100

281670-1G
SIAL281670-1GEA 35.2 EUR
SIAL281670-1G
trans-4-Nitrozimtaldehyd, Sigma-Aldrich®
trans-4-Nitrozimtaldehyd
Formel: C₉H₇NO₃
Molecular Weight: 177.16 g/mol
MDL: MFCD00007379
CAS-Nummer: 49678-08-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR