Mesityloxid, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
4-Methyl-3-penten-2-one
M-Clarity™ Qualitätsstufe = MQ100
281832-100G
SIAL281832-100GEA
57.4
EUR
SIAL281832-100G
Mesityloxid, Sigma-Aldrich®
Mesityloxid
Formel:
C₆H₁₀O Molecular Weight: 98,14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008900 CAS-Nummer: 141-79-7 EINECS: 205-502-5 UN: 1229 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...