Kaliumtrifluoracetat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Trifluoressigsäure Kaliumsalz
M-Clarity™ Qualitätsstufe = MQ200
281883-100G
281883-25G
SIAL281883-100GEA
112
EUR
SIAL281883-100G
SIAL281883-25G
Kaliumtrifluoracetat, Sigma-Aldrich®
Kaliumtrifluoracetat
Formel:
C₂F₃KO₂ Molecular Weight: 152.11 g/mol |
MDL:
MFCD00013215 CAS-Nummer: 2923-16-2 EINECS: 220-877-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...