Bis(trimethylsilyl)methan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

282138-5G
SIAL282138-5GEA 436 EUR
SIAL282138-5G
Bis(trimethylsilyl)methan, Sigma-Aldrich®
Bis(trimethylsilyl)methan
Formel: C₇H₂₀Si₂
Molecular Weight: 160,405 g/mol
MDL: MFCD00008275
CAS-Nummer: 2117-28-4
EINECS: 218-322-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR