Decafluor-4,6-bis(trifluormethyl)cyclohexan, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ100
282316-50G
SIAL282316-50GEA
151
EUR
SIAL282316-50G
Decafluor-4,6-bis(trifluormethyl)cyclohexan, Sigma-Aldrich®
Decafluor-4,6-bis(trifluormethyl)cyclohexan
Formel:
C₈F₁₆ |
MDL:
MFCD00001469 CAS-Nummer: 335-27-3 EINECS: 206-386-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...