Benzolsulfonylfluorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

282332-5G
SIAL282332-5GEA 112 EUR
SIAL282332-5G
Benzolsulfonylfluorid, Sigma-Aldrich®
Benzolsulfonylfluorid
Formel: C₆H₅FO₂S
Molecular Weight: 160,17 g/mol
MDL: MFCD00007412
CAS-Nummer: 368-43-4
EINECS: 206-707-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR