Benzo-15-krone-5 (Benzo-1,4,7,10,13-pentaoxacyclopentadecan), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

282790-5G
SIAL282790-5GEA 126 EUR
SIAL282790-5G
Benzo-15-krone-5 (Benzo-1,4,7,10,13-pentaoxacyclopentadecan), Sigma-Aldrich®
Benzo-15-krone-5 (Benzo-1,4,7,10,13-pentaoxacyclopentadecan)
Formel: C₁₄H₂₀O₅
MDL: MFCD00005945
CAS-Nummer: 14098-44-3
EINECS: 237-947-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR