Graphit

Lieferant: Merck

282863-1KG
SIAL282863-1KGEA 69.5 EUR
SIAL282863-1KG
Graphit
Graphit
Formel: C
Schmelzpunkt: 3827 °C
Dichte: 2,09…2.23 g/cm³ (20 °C)
MDL: MFCD00144065
CAS-Nummer: 7782-42-5
EINECS: 231-955-3
Merck Index: 12,04560

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR