N-Acetyl-ε-caprolactam 99%, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: N-Acetyl-2-oxohexamethyleneimine, N-Acetylcaprolactam

M-Clarity™ Qualitätsstufe = MQ100

283010-250ML
SIAL283010-250MLEA 131 EUR
SIAL283010-250ML
N-Acetyl-ε-caprolactam 99%, Sigma-Aldrich®
N-Acetyl-epsilon-caprolactam

N-Acetylcaprolactam can be used as initiator to investigate kinetics of anionic polymerization of ε-caprolactam, in ring-opening polymerisation of nylon 6.

Formel: C₈H₁₃NO₂
Molecular Weight: 155,20 g/mol
Siedepunkt: 134…135 °C/26 mmHg (Lit.)
Flammpunkt: 122 °C
MDL: MFCD00003262
CAS-Nummer: 1888-91-1
EINECS: 217-565-6

Bestellen

Spezifikation Testergebnisse

Appearance Colourless liquid
Infra-red Spectrum Conforms to structure
Assay Min.98.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR