Ethyl-3-fluorbenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Fluorbenzoesäureethylester

M-Clarity™ Qualitätsstufe = MQ100

283517-10G
SIAL283517-10GEA 85.5 EUR
SIAL283517-10G
Ethyl-3-fluorbenzoat, Sigma-Aldrich®
Ethyl-3-fluorbenzoat
Formel: C₉H₉FO₂
Molecular Weight: 168,17 g/mol
MDL: MFCD00000336
CAS-Nummer: 451-02-5
EINECS: 207-191-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR