Vinylferrocen, Sigma-Aldrich®

Lieferant: Merck


MQ200

283630-1G 283630-5G
SIAL283630-1GEA 121 EUR
SIAL283630-1G SIAL283630-5G
Vinylferrocen, Sigma-Aldrich®
Vinylferrocen
Formel: C₁₂H₁₂Fe
Molecular Weight: 212.07 g/mol
MDL: MFCD00001434
CAS-Nummer: 1271-51-8
EINECS: 215-041-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR